accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

KKCFeCCCCCNNNNNNKKH2OH2OH2O

CAS RN 14459-95-1

IUPAC Name: λ²-iron(2+) tetrapotassium hexakis(iminomethanide) trihydrate
Synonyms: λ²-iron(2+) tetrapotassium hexakis(cyanide) trihydrate
Molecular Weight (g/mol): 422.39
Molecular Formula: C6H6FeK4N6O3
InChi Key: UTYXJYFJPBYDKY-UHFFFAOYSA-N
SMILES: O.O.O.[K+].[K+].[K+].[K+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Quantity 
  • (2)
  • (1)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (11)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (6)
  • (1)
  • (4)
  • (3)
Grade 
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (5)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (11)
  • (1)
  • (3)
  • (3)
  • (6)
  • (1)
  • (4)
  • (3)
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (5)
  • (1)
  • (3)