missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferrocyanide Trihydrate, TRC

Product Code. 30133165
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133165 100 g 100g
30132646 25 g 25g
17987714 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133165 Supplier Toronto Research Chemicals Supplier No. TRCP697305100G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6FeK4N6 . 3(H2O)
Molecular Weight (g/mol) 422.39
Synonym Potassium Ferrocyanide (K4Fe(CN)6) Trihydrate, Potassium Hexacyanoferrate (K4Fe(CN)6) Trihydrate, Potassium Hexacyanoferrate(4-) Trihydrate, Potassium Hexacyanoferrate(II) Trihydrate, Tetrapotassium Ferrocyanide Trihydrate, Tetrapotassium Hexacyanoferrate Trihydrate, Tetrapotassium Hexacyanoferrate(4-) Trihydrate, Tetrapotassium Hexacyanoferrate(II) Trihydrate, ZJ 15 Trihydrate, Potassium Ferrocyanate Trihydrate, Hexacyanoferrate (4-) Tetrapotassium Trihydrate, Hexakis(cyano-C)ferrate Tetrapotassium Trihydrate, Hexakis(cyano-κC)ferrate Tetrapotassium Trihydrate, Iron Potassium Cyanide (FeK4(CN)6) Trihydrate;
IUPAC Name tetrapotassium;iron(2+);hexacyanide;trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].[Fe+2].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N

Specifications

Quantity 100 g
Formula Weight 421.8399
Chemical Name or Material Potassium Ferrocyanide Trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.