accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NON

CAS RN 1453-82-3

IUPAC Name: pyridine-4-carboxamide
Synonyms: isn isonicotineamide I-nicotinamide isoniacinamide
Molecular Weight (g/mol): 122.13
Molecular Formula: C6H6N2O
InChi Key: VFQXVTODMYMSMJ-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (1)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (3)
  • (5)
  • (5)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)