accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ON

CAS RN 14548-46-0

IUPAC Name: 4-benzoylpyridine
Synonyms: 4-benzoylpyridine
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: SKFLCXNDKRUHTA-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=C1)C1=CC=NC=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)