missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Benzoylpyridine, 97%

Product Code. 11459076
Change view
Click to view available options
Quantity:
250g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459076 250g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11459076 Supplier Alfa Aesar™ Supplier No. A17945.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 14548-46-0
Boiling Point 130°C to 131°C (1mmHg)
Flash Point 150°C (302°F)
Molecular Formula C12H9NO
MDL Number MFCD00006430
Quantity 250g
Beilstein 3864
Synonym 4-benzoylpyridine, phenyl pyridin-4-yl methanone, phenyl 4-pyridyl ketone, methanone, phenyl-4-pyridinyl, phenyl 4-pyridinyl methanone, 4-pyridyl phenyl ketone, pyridine, 4-benzoyl, ketone, phenyl 4-pyridyl, gamma-benzoylpyridine, 4-benzoyl pyridine
InChI Key SKFLCXNDKRUHTA-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)C2=CC=NC=C2
IUPAC Name phenyl(pyridin-4-yl)methanone
Molecular Weight (g/mol) 183.21
PubChem CID 26731
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 4-Benzoylpyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.