accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

KCN

CAS RN 151-50-8

IUPAC Name: potassium iminomethanide
Synonyms: potassium cyanide
Molecular Weight (g/mol): 65.12
Molecular Formula: CKN
InChi Key: NNFCIKHAZHQZJG-UHFFFAOYSA-N
SMILES: [K+].[C-]#N
Molecular Weight (g/mol) 
  • (23)
  • (12)
Quantity 
  • (4)
  • (1)
  • (5)
  • (5)
  • (3)
  • (1)
  • (6)
  • (1)
  • (2)
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (7)
  • (12)
  • (3)
  • (2)
Grade 
  • (10)
  • (4)
  • (4)
  • (1)
  • (4)
  • (2)
Physical Form 
  • (4)
  • (4)
  • (4)
  • (6)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (12)
  • (5)
  • (23)
  • (12)
  • (4)
  • (1)
  • (5)
  • (5)
  • (3)
  • (1)
  • (6)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (7)
  • (12)
  • (3)
  • (2)
  • (10)
  • (4)
  • (4)
  • (1)
  • (4)
  • (2)
  • (4)
  • (4)
  • (4)
  • (6)
  • (3)
  • (1)