Product Code. 15653330
Click to view available options
Quantity:
100 g
250 g
1 kg
25 kg
Unit Size:
100g
1kg
250g
25kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
15653330
1 kg
1kg
15643330
100 g
100g
15663330
250 g
250g
15673330
25 kg
25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
15653330
Supplier
Honeywell-Fluka
Supplier No.
312521KG
266.60 EUR valid until 2026-12-31 BEST PRICE on Promo! Use promo code "28038" to get your promotional price.
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥97.0%
Leading the Industry For Over 65 Years Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
CAS
151-50-8
Molecular Formula
CKN
Molecular Weight (g/mol)
65.116
MDL Number
MFCD00011397
InChI Key
NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
PubChem CID
9032
IUPAC Name
potassium;cyanide
SMILES
[C-]#N.[K+]
Specifications
Physical Form
Crystals
Quantity
1 kg
Linear Formula
KCN
UN Number
1680
Beilstein
3593645
Formula Weight
65.12g/mol
Percent Purity
≥97%
Grade
Not Available
Chemical Name or Material
Potassium Cyanide, Reag. Ph. Eur.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?