accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NFCl

CAS RN 15205-15-9

IUPAC Name: 1-(2-chloro-6-fluorophenyl)methanamine
Synonyms: 1-(2-chloro-6-fluorophenyl)methanamine
Molecular Weight (g/mol): 159.59
Molecular Formula: C7H7ClFN
InChi Key: GVULSXIBCHPJEH-UHFFFAOYSA-N
SMILES: NCC1=C(F)C=CC=C1Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results