accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15307-78-5

IUPAC Name: methyl 2-{2-[(2,6-dichlorophenyl)amino]phenyl}acetate
Synonyms: methyl 2-{2-[(2,6-dichlorophenyl)amino]phenyl}acetate
Molecular Weight (g/mol): 310.17
Molecular Formula: C15H13Cl2NO2
InChi Key: VETACGBDFVVKGZ-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=CC=C1NC1=C(Cl)C=CC=C1Cl
Certifications/Compliance 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results