accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3H3COOOCH3CH3H3C

CAS RN 1538-75-6

IUPAC Name: 2,2-dimethylpropanoyl 2,2-dimethylpropanoate
Synonyms: trimethylacetic anhydride
Molecular Weight (g/mol): 186.25
Molecular Formula: C10H18O3
InChi Key: PGZVFRAEAAXREB-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)OC(=O)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)