accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FONNN

CAS RN 1649-18-9

IUPAC Name: 1-(4-fluorophenyl)-4-[4-(pyridin-2-yl)piperazin-1-yl]butan-1-one
Synonyms: azaperone 阿扎哌隆 azaperonum azaperona stresnil(TN) suicalm azaperone janssen brand eucalmyl azaperon janssen brand of azaperone
Molecular Weight (g/mol): 327.4
Molecular Formula: C19H22FN3O
InChi Key: XTKDAFGWCDAMPY-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)CCCN1CCN(CC1)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (2)
  • (1)
Concentration 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (7)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)