CAS RN 1704741-11-6
CAS RN 1704741-11-6
IUPAC Name:
N-[4-({3-cyano-1-[(3,5-dimethyl-1,2-oxazol-4-yl)methyl]-1H-indol-5-yl}oxy)phenyl]pyrrolidine-2-carboxamide
Synonyms:
N-[4-({3-cyano-1-[(3,5-dimethyl-1,2-oxazol-4-yl)methyl]indol-5-yl}oxy)phenyl]pyrrolidine-2-carboxamide
Molecular Weight (g/mol):
455.52
Molecular Formula:
C26H25N5O3
InChi Key:
NDIKFKQBWGMLCA-UHFFFAOYNA-N
SMILES:
CC1=C(CN2C=C(C#N)C3=CC(OC4=CC=C(NC(=O)C5CCCN5)C=C4)=CC=C23)C(C)=NO1
1
–
1
of
1
results
1
Tocris Bioscience™ AZ PFKFB3 67, >98%
CAS: 1704741-11-6 Molecular Formula: C26H25N5O3