accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OFFFFFFFOHCH3H3CCH3OFFFFFFFOHCH3H3CCH3OFFFFFFFOHEu(E/Z)(E/Z)(E/Z)

CAS RN 17631-68-4

IUPAC Name: tris(1,1,1,2,2,3,3-heptafluoro-6-hydroxy-7,7-dimethyloct-5-en-4-one) europium
Synonyms: EuFOD
Molecular Weight (g/mol): 1040.52
Molecular Formula: C30H33EuF21O6
InChi Key: UDXLMYFGTHAWDC-UHFFFAOYSA-N
SMILES: [Eu].CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results