accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1910-42-5

ClClCH3NNCH3

CAS RN 1910-42-5

IUPAC Name: 1,1'-dimethyl-[4,4'-bipyridine]-1,1'-diium dichloride
Synonyms: paraquat dichloride methyl viologen dichloride
Molecular Weight (g/mol): 257.16
Molecular Formula: C12H14Cl2N2
InChi Key: FIKAKWIAUPDISJ-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].C[N+]1=CC=C(C=C1)C1=CC=[N+](C)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)