accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1930-72-9

ONOClNOON

CAS RN 1930-72-9

IUPAC Name: 4-chloro-3,5-dinitrobenzonitrile
Synonyms: 4-chloro-3,5-dinitrobenzonitrile
Molecular Weight (g/mol): 227.56
Molecular Formula: C7H2ClN3O4
InChi Key: SCGDEDHSPCXGEC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC(=C1Cl)[N+]([O-])=O)C#N
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)