CAS RN 202468-31-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
L-Glutamic Acid-13C5,15N, TRC
CAS: 202468-31-3 Molecular Formula: 13C5 H9 15N O4 Molecular Weight (g/mol): 153.086 Synonym: (2S)-2-Aminopentanedioic Acid-13C5,15N IUPAC Name: (2R)-2-(^{15}N)azanyl(1,2,3,4,5-^{13}C_{5})pentanedioic acid SMILES: [15NH2][13C@H]([13CH2][13CH2][13C](=O)O)[13C](=O)O