missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid-13C5,15N, TRC

Product Code. 30012031
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012031 5 mg 5mg
30003365 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012031 Supplier Toronto Research Chemicals Supplier No. TRCG5969625MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202468-31-3
Molecular Formula 13C5 H9 15N O4
Molecular Weight (g/mol) 153.086
Synonym (2S)-2-Aminopentanedioic Acid-13C5,15N
IUPAC Name (2R)-2-(^{15}N)azanyl(1,2,3,4,5-^{13}C_{5})pentanedioic acid
SMILES [15NH2][13C@H]([13CH2][13CH2][13C](=O)O)[13C](=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m1/s1/i1+1,2+1,3+1,4+1,5+1,6+1
Formula Weight 153.067
Chemical Name or Material L-Glutamic Acid-13C5,15N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.