CAS RN 203395-84-0
CAS RN 203395-84-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
OPC 14714, TRC
CAS: 203395-84-0 Molecular Formula: C23 H28 Br N3 O2 Molecular Weight (g/mol): 458.39 Synonym: 2(1H)-Quinolinone, 7-[4-[4-(2-bromophenyl)-1-piperazinyl]butoxy]-3,4-dihydro- (9CI, ACI),7-[4-[4-(2-Bromophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone (ACI),7-[4-[4-(2-bromophenyl)piperazin-1-yl]butoxy]-3,4-dihydroquinolin-2(1H)-one,2-Monobromo Aripiprazole,2-Monobromoaripiprazole IUPAC Name: 7-[4-[4-(2-bromophenyl)piperazin-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one SMILES: Brc1ccccc1N2CCN(CCCCOc3ccc4CCC(=O)Nc4c3)CC2