accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20780-78-3

IOOHN

CAS RN 20780-78-3

IUPAC Name: 7-iodo-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 7-iodo-1H-indole-2,3-dione
Molecular Weight (g/mol): 273.03
Molecular Formula: C8H4INO2
InChi Key: RLGIFVDILDEIHL-UHFFFAOYSA-N
SMILES: IC1=CC=CC2=C1NC(=O)C2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results