accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2ONH2NONN(S)HO(R)HOCH2HN

CAS RN 209216-23-9

IUPAC Name: 2-amino-9-[(3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-6,9-dihydro-3H-purin-6-one hydrate
Synonyms: 2-amino-9-[(3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-3H-purin-6-one hydrate
Molecular Weight (g/mol): 295.3
Molecular Formula: C12H17N5O4
InChi Key: YXPVEXCTPGULBZ-JBDQBEHPSA-N
SMILES: O.NC1=NC(=O)C2=C(N1)N(C=N2)C1C[C@H](O)[C@@H](CO)C1=C
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (5)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)