accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2161-86-6

IUPAC Name: 1-(3-acetyl-2,4,6-trihydroxyphenyl)ethan-1-one
Synonyms: 2,4-diacetylphloroglucinol
Molecular Weight (g/mol): 210.19
Molecular Formula: C10H10O5
InChi Key: PIFFQYJYNWXNGE-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(O)C(C(C)=O)=C(O)C=C1O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)