accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 216143-90-7

CH3OOH3COOCH3

CAS RN 216143-90-7

IUPAC Name: ethyl 5-acetyl-2-ethoxybenzoate
Synonyms: ethyl 5-acetyl-2-ethoxybenzoate
Molecular Weight (g/mol): 236.27
Molecular Formula: C13H16O4
InChi Key: YXLALZDDWWTPFS-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(OCC)C=CC(=C1)C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results