missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-acetyl-2-ethoxybenzoate, 98%, Thermo Scientific™

Product Code. 11343676
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11343676 100 g 100g
11323676 5 g 5g
11333676 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11343676

Brand: Thermo Scientific Alfa Aesar L14750.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216143-90-7
Molecular Formula C13H16O4
Molecular Weight (g/mol) 236.27
MDL Number MFCD01075665
InChI Key YXLALZDDWWTPFS-UHFFFAOYSA-N
PubChem CID 5051139
IUPAC Name ethyl 5-acetyl-2-ethoxybenzoate
SMILES CCOC(=O)C1=C(OCC)C=CC(=C1)C(C)=O

Specifications

Melting Point 51°C to 54°C
Quantity 100 g
Synonym ethyl5-acetyl-2-ethoxybenzoate, acmc-1cq98, ethyl 5-acetyl-2-ethoxybenzoate, 5-acetyl-2-ethoxybenzoic acid ethyl ester, 5-acetyl-2-ethoxy-benzoic acid ethyl ester, benzoic acid,5-acetyl-2-ethoxy-, ethyl ester
Formula Weight 236.27
Percent Purity 98%
Chemical Name or Material Ethyl 5-acetyl-2-ethoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.