accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CO(E)(E)

CAS RN 21662-16-8

IUPAC Name: (2E,4E)-dodeca-2,4-dienal
Synonyms: (2E,4E)-dodeca-2,4-dienal
Molecular Weight (g/mol): 180.29
Molecular Formula: C12H20O
InChi Key: QKTZBZWNADPFOL-BNFZFUHLSA-N
SMILES: CCCCCCC\C=C\C=C\C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results