CAS RN 223128-24-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(2-Pyridinyl)benzoyl Chloride Hydrochloride, TRC
CAS: 223128-24-3 Molecular Formula: C12H8ClNO Molecular Weight (g/mol): 254.11 Synonym: 4-(Pyridin-2-yl)benzoyl Chloride IUPAC Name: 4-pyridin-2-ylbenzoyl chloride;hydrochloride SMILES: Cl.ClC(=O)c1ccc(cc1)c2ccccn2