accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OO(S)NHO(S)NH2OHO

CAS RN 22839-47-0

IUPAC Name: 3-amino-3-[(1-methoxy-1-oxo-3-phenylpropan-2-yl)carbamoyl]propanoic acid
Synonyms: aminosweet
Molecular Weight (g/mol): 294.31
Molecular Formula: C14H18N2O5
InChi Key: IAOZJIPTCAWIRG-UHFFFAOYNA-N
SMILES: COC(=O)C(CC1=CC=CC=C1)NC(=O)C(N)CC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)