missing translation for 'onlineSavingsMsg'
Learn More

Aspartame, TRC

Product Code. 17967564
Change view
Click to view available options
Quantity:
25 mg
1 g
5 g
Unit Size:
1g
25mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17967564 5 g 5g
17937664 25 mg 25mg
17937104 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17967564 Supplier Toronto Research Chemicals Supplier No. TRCA7900155G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22839-47-0
Synonym L-Phenylalanine, N-L-α-aspartyl-, 1-methyl ester∣Succinamic acid, 3-amino-N-(α-carboxyphenethyl)-, N-methyl ester, stereoisomer (8CI)∣Aspartame∣Aspartylphenylalanine methyl ester∣Canderel∣Dipeptide sweetener∣E 951∣Equal∣Finn∣L-Aspartame∣L-Aspartyl-L-3-phenylalanine methyl ester∣L-Aspartyl-L-phenylalanine methyl ester∣L-Aspartyl-L-phenylalanyl methyl ester∣L-α-Aspartyl-L-phenylalanine methyl ester∣Methyl aspartylphenylalanate∣NutraSweet∣Pal Sweet∣Pal Sweet Diet G 100∣Palsweet Diet∣Sladex∣Sweet dipeptide∣Zero-Cal∣α-Aspartame∣α-L-Aspartyl-L-phenylalanine methyl ester∣α-Sweet

Specifications

Format Neat
Molecular Weight (g/mol) 294.3
Product Type API
Quantity 5 g
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.