CAS RN 2362-10-9
IUPAC Name:
(chloromethyl)({[(chloromethyl)dimethylsilyl]oxy})dimethylsilane
Synonyms:
(chloromethyl)({[(chloromethyl)dimethylsilyl]oxy})dimethylsilane
Molecular Weight (g/mol):
231.26
Molecular Formula:
C6H16Cl2OSi2
InChi Key:
NBGGEWGFZUDQKZ-UHFFFAOYSA-N
SMILES:
C[Si](C)(CCl)O[Si](C)(C)CCl
1
–
1
of
1
results
1
1,3-Bis(chloromethyl)tetramethyldisiloxane, 97%, Thermo Scientific™
CAS: 2362-10-9 MDL Number: MFCD00000877