accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OCH3OOOHCH3HOO(E)

CAS RN 24280-93-1

IUPAC Name: (4E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydro-2-benzofuran-5-yl)-4-methylhex-4-enoic acid
Synonyms: mycophenolic acid mycophenoic acid 麦考酚酸 mycophenolic acid (substance) acide mycophenolique acido micofenolico mycophenolic acid (product) acidum mycophenolicum micofenolico acido acid, mycophenolic
Molecular Weight (g/mol): 320.34
Molecular Formula: C17H20O6
InChi Key: HPNSFSBZBAHARI-RUDMXATFSA-N
SMILES: COC1=C(C)C2=C(C(=O)OC2)C(O)=C1C\C=C(/C)CCC(O)=O
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (6)
Physical Form 
  • (1)
  • (3)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (8)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)