accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOO

CAS RN 25333-24-8

IUPAC Name: methyl 4-oxo-4-phenylbutanoate
Synonyms: methyl 4-oxo-4-phenylbutanoate
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: XVRCVKWYKYJEIG-UHFFFAOYSA-N
SMILES: COC(=O)CCC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)