missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-benzoylpropionate, 98%

Product Code. 11494387
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494387 25 g 25g
11484387 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494387 Supplier Thermo Scientific Alfa Aesar Supplier No. B20435.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25333-24-8 | C11H12O3 | 192.214 g/mol

Methyl 3-benzoylpropionate is used as a chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-benzoylpropionate is used as a chemical and organic intermediate.

Solubility
Soluble in Chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25333-24-8
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00014906
InChI Key XVRCVKWYKYJEIG-UHFFFAOYSA-N
Synonym methyl 3-benzoylpropionate, 3-benzoylpropionic acid methyl ester, propionic acid, 3-benzoyl-, methyl ester, methyl 4-oxo-4-phenylbutyrate, benzenebutanoic acid, gamma-oxo-, methyl ester, benzenebutanoic acid, .gamma.-oxo-, methyl ester, benzenebutanoic acid, g-oxo-, methyl ester, acmc-209gja, methyl3-benzoylpropionate, methyl-3-benzoylpropionate
PubChem CID 141190
IUPAC Name methyl 4-oxo-4-phenylbutanoate
SMILES COC(=O)CCC(=O)C1=CC=CC=C1

Specifications

Boiling Point 172°C to 174°C (10 mmHg)
Refractive Index 1.524
Quantity 25 g
Beilstein 2211627
Solubility Information Soluble in Chloroform.
Formula Weight 192.22
Percent Purity 98%
Chemical Name or Material Methyl 3-benzoylpropionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.