accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27329-70-0

HOBOHOO

CAS RN 27329-70-0

IUPAC Name: (5-formylfuran-2-yl)boronic acid
Synonyms: 5-formylfuran-2-ylboronic acid
Molecular Weight (g/mol): 139.9
Molecular Formula: C5H5BO4
InChi Key: JUWYQISLQJRRNT-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(O1)C=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)