missing translation for 'onlineSavingsMsg'
Learn More

(5-Formylfuran-2-yl)boronic Acid, TRC

Product Code. 30180276
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180276 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30180276 Supplier Toronto Research Chemicals Supplier No. TRCF7003455G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 27329-70-0
Molecular Formula C5 H5 B O4
Quantity 5 g
Synonym Boronic acid, B-(5-formyl-2-furanyl)-, 2-Furanboronic acid, 5-formyl- (8CI), Boronic acid, (5-formyl-2-furanyl)- (9CI), B-(5-Formyl-2-furanyl)boronic acid, (2-Formylfuran-5-yl)boronic acid, (5-Formylfuran-2-yl)boronic acid, 2-Formyl-5-furanboronic acid, 2-Formylfuran-5-boric acid, 5-Formyl-2-furanboronic acid, 5-Formyl-2-furylboronic acid, Lapatinib related, AH23757X, GR82029X, GR82029X Test Mix 1, Lapatinib related
InChI Formula InChI=1S/C5H5BO4/c7-3-4-1-2-5(10-4)6(8)9/h1-3,8-9H
SMILES OB(O)c1oc(C=O)cc1
IUPAC Name (5-formylfuran-2-yl)boronic acid
Molecular Weight (g/mol) 139.9
Formula Weight 140.0281
Chemical Name or Material 5-Formyl-2-furanylboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.