accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27489-62-9

IUPAC Name: 4-aminocyclohexan-1-ol
Synonyms: 4-aminocyclohexan-1-ol
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: IMLXLGZJLAOKJN-UHFFFAOYSA-N
SMILES: NC1CCC(O)CC1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)