missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Aminocyclohexanol, TRC

Product Code. 30005806
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005806 5 g 5g
30005575 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005806 Supplier Toronto Research Chemicals Supplier No. TRCA6039955G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27489-62-9
Molecular Formula C6 H13 N O
Molecular Weight (g/mol) 115.174
Synonym Cyclohexanol, 4-amino-, trans-, 4-trans-Aminocyclohexanol, 4-trans-Hydroxycyclohexylamine, trans-1,4-Cyclohexanolamine, trans-1-Amino-4-cyclohexanol, trans-1-Amino-4-hydroxycyclohexane, trans-4-Amino-1-cyclohexanol, trans-4-Hydroxycyclohexylamine
IUPAC Name 4-aminocyclohexanol
SMILES N[C@@H]1CC[C@@H](O)CC1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H13NO/c7-5-1-3-6(8)4-2-5/h5-6,8H,1-4,7H2/t5-,6-
Formula Weight 115.0997
Chemical Name or Material trans-4-Aminocyclohexanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.