accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOClCl

CAS RN 2905-60-4

IUPAC Name: 2,3-dichlorobenzoyl chloride
Synonyms: benzoyl chloride, dichloro-
Molecular Weight (g/mol): 209.45
Molecular Formula: C7H3Cl3O
InChi Key: YBONBWJSFMTXLE-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CC(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)