missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzoyl chloride, 98%

Product Code. 11337466
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337466 100 g 100g
11317466 5 g 5g
11327466 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11337466 Supplier Thermo Scientific Alfa Aesar Supplier No. L09506.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2905-60-4 | C7H3Cl3O | 209.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-60-4
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00035937
InChI Key YBONBWJSFMTXLE-UHFFFAOYSA-N
Synonym benzoyl chloride, dichloro, dichlorobenzoyl chloride, benzoyl chloride, 2,3-dichloro, unii-i50f21ps1n, 2,3-dichlorobenzoylchloride, acmc-1cfin, 2,3-dichlorobenzoyl-chloride, ksc202s8t, 2, 3-dichlorobenzoyl chloride, 2,3-dcoc
PubChem CID 17944
IUPAC Name 2,3-dichlorobenzoyl chloride
SMILES ClC(=O)C1=CC=CC(Cl)=C1Cl

Specifications

Melting Point 28°C to 32°C
Boiling Point 140°C (14 mmHg)
Flash Point 167°C (332°F)
Odor Pungent
Quantity 100 g
UN Number UN3261
Beilstein 2575973
Sensitivity Moisture sensitive
Formula Weight 209.46
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorobenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.