accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH

CAS RN 2928-43-0

IUPAC Name: {[1,1'-biphenyl]-2-yl}methanol
Synonyms: [1,1'-biphenyl]-2-ylmethanol
Molecular Weight (g/mol): 184.24
Molecular Formula: C13H12O
InChi Key: VKTQADPEPIVMHK-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)