accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COFOCH3

CAS RN 295779-86-1

IUPAC Name: 1-(3-fluoro-2-methoxyphenyl)ethan-1-one
Synonyms: 1-(3-fluoro-2-methoxyphenyl)ethanone
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: XWKGHXBPAPLVPW-UHFFFAOYSA-N
SMILES: COC1=C(C=CC=C1F)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results