accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CNOH

CAS RN 29767-97-3

IUPAC Name: 2-methyl-4-(pyridin-2-yl)but-3-yn-2-ol
Synonyms: 2-methyl-4-(pyridin-2-yl)but-3-yn-2-ol
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: YBUWIVMOMHEALF-UHFFFAOYSA-N
SMILES: CC(C)(O)C#CC1=CC=CC=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results