accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNH(S)(R)H2N(S)OH(R)O(S)O(R)OH(S)OH(S)OO(R)(S)H2NOH(R)OH(R)OH(R)HOO(S)(R)OH

CAS RN 31282-04-9

IUPAC Name: (3'R,3aS,4S,4'R,5'R,6R,6'R,7S,7aS)-4-{[(1R,2S,3R,5S,6R)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy}-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)-tetrahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
Synonyms: hygromycin B
Molecular Weight (g/mol): 527.52
Molecular Formula: C20H37N3O13
InChi Key: GRRNUXAQVGOGFE-KPBUCVLVSA-N
SMILES: CN[C@H]1C[C@@H](N)[C@H](O)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@@H]3OC4(O[C@H]23)O[C@H]([C@@H](N)CO)[C@H](O)[C@@H](O)[C@H]4O)[C@@H]1O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
Color 
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)
Type 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)