accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 32809-16-8

IUPAC Name: 3-(3,5-dichlorophenyl)-1,5-dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione
Synonyms: procymidone sumilex dicyclidine sumisclex
Molecular Weight (g/mol): 284.14
Molecular Formula: C13H11Cl2NO2
InChi Key: QXJKBPAVAHBARF-UHFFFAOYNA-N
SMILES: CC12CC1(C)C(=O)N(C2=O)C1=CC(Cl)=CC(Cl)=C1
Molecular Weight (g/mol) 
  • (7)
  • (4)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)
  • (1)
Concentration 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (1)
  • (3)
  • (7)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)