missing translation for 'onlineSavingsMsg'
Learn More

Procymidox, TRC

Product Code. 30427640
Change view
Click to view available options
Quantity:
50 mg
250 mg
5 g
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427640 5 g -
30409295 50 mg 50mg
30418264 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427640 Supplier Toronto Research Chemicals Supplier No. TRCP7558555G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32809-16-8
Molecular Formula C13 H11 Cl2 N O2
Molecular Weight (g/mol) 284.14
Synonym Sumilex, 3-(3,5-dichlorophenyl)-1,5-dimethyl-1H-pyrrole-2,5-dione
IUPAC Name 3-(3,5-dichlorophenyl)-1,5-dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione
SMILES CC12CC1(C)C(=O)N(C2=O)c3cc(Cl)cc(Cl)c3

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H11Cl2NO2/c1-12-6-13(12,2)11(18)16(10(12)17)9-4-7(14)3-8(15)5-9/h3-5H,6H2,1-2H3
Formula Weight 283.0167
Chemical Name or Material Procymidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.