accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHClF

CAS RN 348-62-9

IUPAC Name: 4-chloro-2-fluorophenol
Synonyms: 4-chloro-2-fluorophenol
Molecular Weight (g/mol): 146.55
Molecular Formula: C6H4ClFO
InChi Key: ZKMUKBBWORLNLA-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)