accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3697-24-3

IUPAC Name: 5-methylchrysene
Synonyms: 5-methylchrysene
Molecular Weight (g/mol): 242.32
Molecular Formula: C19H14
InChi Key: GOHBXWHNJHENRX-UHFFFAOYSA-N
SMILES: CC1=CC2=CC=CC=C2C2=C1C1=CC=CC=C1C=C2
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (1)