missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl Chrysene, TRC

Product Code. 30040905
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040905 5 mg 5mg
30040563 50 mg 50mg
18090060 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040905 Supplier Toronto Research Chemicals Supplier No. TRCM2651355MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3697-24-3
Molecular Formula C19 H14
Molecular Weight (g/mol) 242.31
Synonym Chrysene, 5-methyl-, 5-Methylchrysene, NSC 407620
IUPAC Name 5-methylchrysene
SMILES Cc1cc2ccccc2c3ccc4ccccc4c13

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H14/c1-13-12-15-7-3-4-8-16(15)18-11-10-14-6-2-5-9-17(14)19(13)18/h2-12H,1H3
Formula Weight 242.1096
Percent Purity >95%
Chemical Name or Material 5-Methylchrysene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.