accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOFFF

CAS RN 372-31-6

IUPAC Name: ethyl 4,4,4-trifluoro-3-oxobutanoate
Synonyms: ethyl 4,4,4-trifluoro-3-oxobutanoate
Molecular Weight (g/mol): 184.11
Molecular Formula: C6H7F3O3
InChi Key: OCJKUQIPRNZDTK-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)