accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONSNCH3

CAS RN 383146-88-1

IUPAC Name: ethyl 4-methyl-2-(pyridin-2-yl)-1,3-thiazole-5-carboxylate
Synonyms: ethyl 4-methyl-2-(pyridin-2-yl)-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 248.3
Molecular Formula: C12H12N2O2S
InChi Key: UGCVLLXXBVTJPK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N=C(S1)C1=CC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results