accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 403-15-6

CH3FOHO

CAS RN 403-15-6

IUPAC Name: 4-fluoro-3-methylbenzoic acid
Synonyms: 4-fluoro-3-methylbenzoic acid
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: VOCCEVKUXUIHOI-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results