accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClCH3CH3NH2

CAS RN 41100-52-1

IUPAC Name: hydrogen 3,5-dimethyladamantan-1-amine chloride
Synonyms: hydrogen memantine chloride
Molecular Weight (g/mol): 215.77
Molecular Formula: C12H22ClN
InChi Key: LDDHMLJTFXJGPI-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)